Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-4-(4-isopropylanilino)tetrahydro-1(2H)-pyridinecarboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

045162-500MG 045162-1G 045162-5G
101869-448EA 181.54 USD InStock
101869-448 101869-450 101869-452
tert-Butyl-4-(4-isopropylanilino)tetrahydro-1(2H)-pyridinecarboxylate ≥95%
tert-Butyl-4-(4-isopropylanilino)tetrahydro-1(2H)-pyridinecarboxylate
Hazard: Irritant


Formula: C₁₉H₃₀N₂O₂
Melting Pt: 99…100 °C
MDL Number: MFCD06659568
CAS Number: 241499-44-5
Order Now
Specification Test Results
Melting point 99-100°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty