Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-isopropylphenyl)propanoic acid hydrate ≥95%

Supplier: MATRIX SCIENTIFIC MS

045191-500MG 045191-1G 045191-5G
101869-606EA 375.38 USD InStock
101869-606 101869-608 101869-610
3-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-isopropylphenyl)propanoic acid hydrate ≥95%
3-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-isopropylphenyl)propanoic acid hydrate
Hazard: Irritant


Formula: C₂₇H₂₉NO₅
MW: 447.54 g/mol
Melting Pt: 183…185 °C
MDL Number: MFCD06659576
CAS Number: 1049746-57-7
Order Now
Specification Test Results
Melting point 183-185°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty