Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-6-cyanoimidazo[1,2-a]pyridine-2-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

045205-500MG 045205-1G 045205-5G
101869-678EA 183.08 USD InStock
101869-678 101869-680 101869-682
Ethyl-6-cyanoimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 6-cyanoimidazo[1,2-a]pyridine-2-carboxylate
Hazard: Irritant


Formula: C₁₁H₉N₃O₂
Melting Pt: 177-180°C
MDL Number: MFCD06739178
CAS Number: 214958-33-5
Order Now
Specification Test Results
Melting point 177-180°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty