Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-[(3-chlorobenzoyl)amino]-6-methyl-2-oxo-2H-pyran-5-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

045238-500MG 045238-1G 045238-5G
101869-858EA 375.38 USD InStock
101869-858 101869-860 101869-862
Methyl-3-[(3-chlorobenzoyl)amino]-6-methyl-2-oxo-2H-pyran-5-carboxylate ≥95%
Methyl-3-[(3-chlorobenzoyl)amino]-6-methyl-2-oxo-2H-pyran-5-carboxylate
Hazard: Irritant


Formula: C₁₅H₁₂ClNO₅
MW: 321.72 g/mol
Melting Pt: 171…172 °C
MDL Number: MFCD00139846
CAS Number: 339009-28-8
Order Now
Specification Test Results
Melting point 171-172°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty