Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-{[(2,6-Dichlorobenzyl)sulfanyl]methyl}-4-ethyl-4H-1,2,4-triazole-3-thiol ≥95%

Supplier: MATRIX SCIENTIFIC MS

045266-500MG 045266-1G 045266-5G
101870-014EA 747.69 USD InStock
101870-014 101870-016 101870-018
5-{[(2,6-Dichlorobenzyl)sulfanyl]methyl}-4-ethyl-4H-1,2,4-triazole-3-thiol ≥95%
5-{[(2,6-Dichlorobenzyl)sulfanyl]methyl}-4-ethyl-4H-1,2,4-triazole-3-thiol
Hazard: Irritant


Formula: C₁₂H₁₃Cl₂N₃S₂
Melting Pt: 175…178 °C
MDL Number: MFCD00244410
CAS Number: 344269-70-1
Order Now
Specification Test Results
Melting point 175-178°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty