Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4-Methylphenyl)-2,1-benzisoxazole-5-carbaldehyde ≥95%

Supplier: MATRIX SCIENTIFIC MS

045270-500MG 045270-1G 045270-5G
101870-038EA 747.69 USD InStock
101870-038 101870-040 101870-042
3-(4-Methylphenyl)-2,1-benzisoxazole-5-carbaldehyde ≥95%
3-(4-Methylphenyl)-2,1-benzisoxazole-5-carbaldehyde
Hazard: Irritant


Formula: C₁₅H₁₁NO₂
Melting Pt: 152…154 °C
MDL Number: MFCD00663843
CAS Number: 328547-38-2
Order Now
Specification Test Results
Melting point 152-154°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty