Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Benzyl-5-bromo-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

045469-500MG 045469-1G 045469-5G
101870-986EA 500 USD InStock
101870-986 101870-988 101870-990
1-Benzyl-5-bromo-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%
1-Benzyl-5-bromo-2-oxo-1,2-dihydro-3-pyridine carboxylic acid
Hazard: Irritant


Formula: C₁₃H₁₀BrNO₃
MW: 308.14 g/mol
Melting Pt: 233…235 °C
MDL Number: MFCD05663803
CAS Number: 101384-63-8
Order Now
Specification Test Results
Melting point 165-168°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty