Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Bromophenyl)-4-(2-hydroxyethyl)-5-methyl-1,2-dihydro-3H-pyrazol-3-one ≥95%

Supplier: MATRIX SCIENTIFIC MS

045492-500MG 045492-1G 045492-5G
101871-122EA 278.46 USD InStock
101871-122 101871-124 101871-126
2-(4-Bromophenyl)-4-(2-hydroxyethyl)-5-methyl-1,2-dihydro-3H-pyrazol-3-one ≥95%
2-(4-Bromophenyl)-4-(2-hydroxyethyl)-5-methyl-1,2-dihydro-3H-pyrazol-3-one
Hazard: Irritant


Formula: C₁₂H₁₃BrN₂O₂
Melting Pt: 171…173 °C
MDL Number: MFCD06659582
CAS Number: 885949-90-6
Order Now
Specification Test Results
Melting point 171-173°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty