Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-[2-nitro-4-(trifluoromethyl)anilino]propanoate ≥95%

Supplier: MATRIX SCIENTIFIC MS

045511-500MG 045511-1G 045511-5G
101871-220EA 181.54 USD InStock
101871-220 101871-222 101871-224
Methyl-3-[2-nitro-4-(trifluoromethyl)anilino]propanoate ≥95%
Methyl-3-[2-nitro-4-(trifluoromethyl)anilino]propanoate
Hazard: Irritant


Formula: C₁₁H₁₁F₃N₂O₄
Melting Pt: 85…88 °C
MDL Number: MFCD06659588
CAS Number: 885949-93-9
Order Now
Specification Test Results
Melting point 85-88°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty