Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-[2-Nitro-4-(trifluoromethyl)anilino]propanoic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

045512-500MG 045512-1G 045512-5G
101871-226EA 278.46 USD InStock
101871-226 101871-228 101871-230
3-[2-Nitro-4-(trifluoromethyl)anilino]propanoic acid ≥95%
3-[2-Nitro-4-(trifluoromethyl)anilino]propanoic acid
Hazard: Irritant


Formula: C₁₀H₉F₃N₂O₄
Melting Pt: 128…131 °C
MDL Number: MFCD01240395
CAS Number: 37040-43-0
Order Now
Specification Test Results
Melting point 128-131°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty