Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-5-bromo-2-methoxy-3-methylbenzenecarboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

045514-500MG 045514-1G 045514-5G
101871-238EA 353.85 USD InStock
101871-238 101871-240 101871-242
Methyl-5-bromo-2-methoxy-3-methylbenzenecarboxylate ≥95%
Methyl-5-bromo-2-methoxy-3-methylbenzenecarboxylate
Hazard: Irritant


Formula: C₁₀H₁₁BrO₃
Melting Pt: 60…62 °C
MDL Number: MFCD06659589
CAS Number: 722497-32-7
Order Now
Specification Test Results
Melting point 60-62°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty