Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-[(4-Chlorophenyl)methyl]-1,3,4-oxadiazol-2-ol ≥95%

Supplier: MATRIX SCIENTIFIC MS

058493-500MG 058493-1G
101914-910EA 315.38 USD InStock
101914-910 101914-912
5-[(4-Chlorophenyl)methyl]-1,3,4-oxadiazol-2-ol ≥95%
5-[(4-Chlorophenyl)methyl]-1,3,4-oxadiazol-2-ol
Hazard: Irritant


Formula: C₉H₇ClN₂O₂
Melting Pt: 112…114 °C
MDL Number: MFCD17392925
CAS Number: 34547-04-1
Order Now
Specification Test Results
Melting point 112-114°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty