Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4-hydroxy-2-(pyridin-4-yl)pyrimidine-5-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

058651-500MG 058651-1G 058651-5G
101915-244EA 295.38 USD InStock
101915-244 101915-246 101915-248
Ethyl-4-hydroxy-2-(pyridin-4-yl)pyrimidine-5-carboxylate ≥95%
Ethyl-4-hydroxy-2-(pyridin-4-yl)pyrimidine-5-carboxylate
Hazard: Irritant


Formula: C₁₂H₁₁N₃O₃
MW: 245.24 g/mol
Melting Pt: 198…200 °C
MDL Number: MFCD11047091
CAS Number: 56406-44-1
Order Now
Specification Test Results
Melting point 198-200°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty