Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-5-nitro-2,3-dihydro-1H-1,3-benzodiazol-2-one ≥95%

Supplier: MATRIX SCIENTIFIC MS

058679-500MG 058679-1G
101915-372EA 598.46 USD InStock
101915-372 101915-374
1-Methyl-5-nitro-2,3-dihydro-1H-1,3-benzodiazol-2-one ≥95%
1-Methyl-5-nitro-2,3-dihydro-1H-1,3-benzodiazol-2-one
Hazard: Irritant


Formula: C₈H₇N₃O₃
Melting Pt: 300…302 °C
MDL Number: MFCD18157683
CAS Number: 66108-85-8
Order Now
Specification Test Results
Melting point 300-302°C

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty