Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4-Diphenylpiperidine hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

008692-1G 008692-5G
101795-186EA 80 USD InStock
101795-186 101795-188
4,4-Diphenylpiperidine hydrochloride ≥95%
4,4-Diphenylpiperidine hydrochloride
Hazard: Irritant


Formula: C₁₇H₂₀ClN
Melting Pt: 300…305 °C
MDL Number: MFCD01700799
CAS Number: 63675-71-8
Order Now
Specification Test Results
Melting point 300-305°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty