Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Acetylpiperidine-4-carboxylic acid ≥98%

Supplier: MATRIX SCIENTIFIC MS

008731-25G 008731-100G
101795-308EA 24.62 USD InStock
101795-308 101795-310
1-Acetylpiperidine-4-carboxylic acid ≥98%
1-Acetylpiperidine-4-carboxylic acid
Hazard: Irritant


Formula: C₈H₁₃NO₃
Melting Pt: 163…178 °C
MDL Number: MFCD00023706
CAS Number: 25503-90-6
Order Now
Specification Test Results
Melting point 180-183°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty