Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethyl)salicylic acid ≥98%

Supplier: MATRIX SCIENTIFIC MS

008787-25G 008787-100G
101795-456EA 53.85 USD InStock
101795-456 101795-458
4-(Trifluoromethyl)salicylic acid ≥98%
4-(Trifluoromethyl)salicylic acid
Hazard: Irritant


Formula: C₈H₅F₃O₃
Melting Pt: 176-178°C
MDL Number: MFCD01529657
CAS Number: 328-90-5
Order Now
Specification Test Results
Melting point 176-178°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty