Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9-Fluorenylmethyl pentafluorophenyl carbonate ≥98%

Supplier: MATRIX SCIENTIFIC MS

008938-1G 008938-5G
101795-860EA 80 USD InStock
101795-860 101795-862
9-Fluorenylmethyl pentafluorophenyl carbonate ≥98%
9-Fluorenylmethyl pentafluorophenyl carbonate
Hazard: Irritant


Formula: C₂₁H₁₁F₅O₃
Melting Pt: 85…87 °C
MDL Number: MFCD00013261
CAS Number: 88744-04-1
Order Now
Specification Test Results
Melting point 85-87°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty