Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-(Hexafluoroisopropylidene)diphthalic anhydride ≥99%

Supplier: MATRIX SCIENTIFIC MS

002952-25G 002952-100G
101784-966EA 90.77 USD InStock
101784-966 102550-826
4,4'-(Hexafluoroisopropylidene)diphthalic anhydride ≥99%
4,4'-(Hexafluoroisopropylidene)diphthalic anhydride
TCA listed: T
Hazard: Corrosive/Moisture Sensitive


Formula: C₈H₄O₃
Melting Pt: 244…247 °C
MDL Number: MFCD00039143
CAS Number: 1107-00-2
Order Now
Specification Test Results
Melting point 241-243°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty