Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxybenzoic anhydride ≥98%

Supplier: MATRIX SCIENTIFIC MS

003018-5G 003018-25G
101785-090EA 130.77 USD InStock
101785-090 101785-092
4-Methoxybenzoic anhydride ≥98%
4-Methoxybenzoic anhydride
Hazard: Irritant/Moisture Sensitive


Formula: C₁₆H₁₄O₅
MW: 286.29 g/mol
Melting Pt: 96…98 °C
MDL Number: MFCD00017175
CAS Number: 794-94-5
Order Now
Specification Test Results
Melting point 96-98°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty