Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diphenylmethane-4,4'-dicarboxylic acid ≥97%

Supplier: MATRIX SCIENTIFIC MS

003369-1G 003369-5G
101785-626EA 96.92 USD InStock
101785-626 101785-628
Diphenylmethane-4,4'-dicarboxylic acid ≥97%
Diphenylmethane-4,4'-dicarboxylic acid
Hazard: Irritant


Formula: C₁₅H₁₂O₄
Melting Pt: 346…350 °C
MDL Number: MFCD00060322
CAS Number: 790-83-0
Order Now
Specification Test Results
Melting point 346-350°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty