Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,6,7,8-Tetrafluorobenzo-1,4-dioxene ≥97%

Supplier: MATRIX SCIENTIFIC MS

003461-1G 003461-5G
101785-732EA 80 USD InStock
101785-732 101785-734
5,6,7,8-Tetrafluorobenzo-1,4-dioxene ≥97%
5,6,7,8-Tetrafluorobenzo-1,4-dioxene
Hazard: Irritant


Formula: C₈H₄F₄O₂
Melting Pt: 78…79 °C
MDL Number: MFCD00229993
CAS Number: 1743-87-9
Order Now
Specification Test Results
Melting point 78-79°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty