Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-[4-(Trifluoromethyl)benzoyl]-L-methioninemethyl ester

Supplier: MATRIX SCIENTIFIC MS

003732-1G 003732-5G
101786-124EA 80 USD InStock
101786-124 101786-126
N-[4-(Trifluoromethyl)benzoyl]-L-methioninemethyl ester
N-[4-(Trifluoromethyl)benzoyl]-L-methioninemethyl ester
Hazard: Irritant


Formula: C₁₄H₁₆F₃NO₃S
Melting Pt: 94…96 °C
MDL Number: MFCD00085178
CAS Number: 175202-25-2
Order Now
Specification Test Results
Melting point 94-96°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty