Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Diamino-3-fluoro-4-(trifluoromethyl)pyridine ≥98%

Supplier: MATRIX SCIENTIFIC MS

021139-250MG 021139-1G
101811-428EA 186.15 USD InStock
101811-428 101811-430
2,6-Diamino-3-fluoro-4-(trifluoromethyl)pyridine ≥98%
2,6-Diamino-3-fluoro-4-(trifluoromethyl)pyridine
Hazard: Irritant


Formula: C₆H₅F₄N₃
MW: 195.12 g/mol
Melting Pt: 70…71 °C
MDL Number: MFCD08687525
CAS Number: 737000-87-2
Order Now
Specification Test Results
Melting point 70-71°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty