Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-2-methoxypyridine ≥98%

Supplier: MATRIX SCIENTIFIC MS

023787-25G 023787-100G
101816-746EA 75.38 USD InStock
101816-746 102551-036
4-Amino-2-methoxypyridine ≥98%
4-Amino-2-methoxypyridine
Hazard: Irritant


Formula: C₆H₈N₂O
MW: 124.1400 g/mol
Melting Pt: 87…88 °C
MDL Number: MFCD06738657
CAS Number: 20265-39-8
Order Now
Specification Test Results
Melting point 87-88°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty