Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Ethyl-5,7-dimethyl[1,2,4]triazolo[1,5-a]pyrimidin-2-amine ≥95%

Supplier: MATRIX SCIENTIFIC MS

046687-500MG 046687-1G
101876-540EA 278.46 USD InStock
101876-540 101876-542
6-Ethyl-5,7-dimethyl[1,2,4]triazolo[1,5-a]pyrimidin-2-amine ≥95%
6-Ethyl-5,7-dimethyl[1,2,4]triazolo[1,5-a]pyrimidin-2-amine
Hazard: Irritant


Formula: C₉H₁₃N₅
Melting Pt: 297…300 °C
MDL Number: MFCD07402021
CAS Number: 924868-96-2
Order Now
Specification Test Results
Melting point 297-300°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty