Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4-hydroxy-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

046696-500MG 046696-1G 046696-5G
101876-592EA 278.46 USD InStock
101876-592 101876-594 101876-596
Ethyl-4-hydroxy-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate ≥95%
Ethyl-4-hydroxy-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate
Hazard: Irritant


Formula: C₁₃H₁₀F₃NO₃S
Melting Pt: 118…121 °C
MDL Number: MFCD00816788
CAS Number: 227199-08-8
Order Now
Specification Test Results
Melting point 118-121°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty