Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-N-[3-(1-pyrrolidinyl)propyl]benzenesulfonamide ≥95%

Supplier: MATRIX SCIENTIFIC MS

046769-500MG 046769-1G
101876-934EA 181.54 USD InStock
101876-934 101876-936
4-Chloro-N-[3-(1-pyrrolidinyl)propyl]benzenesulfonamide ≥95%
4-Chloro-N-[3-(1-pyrrolidinyl)propyl]benzenesulfonamide
Hazard: Irritant


Formula: C₁₃H₁₉ClN₂O₂S
Melting Pt: 52…54 °C
MDL Number: MFCD02557035
CAS Number: 321718-12-1
Order Now
Specification Test Results
Melting point 52-54°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty