Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(1-Benzyl-4-piperidinyl)-2-phenyl-6-oxo-3-piperidinecarboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

046991-500MG 046991-1G 046991-5G
101877-932EA 375.38 USD InStock
101877-932 101877-934 101877-936
1-(1-Benzyl-4-piperidinyl)-2-phenyl-6-oxo-3-piperidinecarboxylic acid ≥95%
1-(1-Benzyl-4-piperidinyl)-2-phenyl-6-oxo-3-piperidinecarboxylic acid
Hazard: Irritant


Formula: C₂₄H₂₈N₂O₃
Melting Pt: 214…216 °C
MDL Number: MFCD09027092
CAS Number: 937604-14-3
Order Now
Specification Test Results
Melting point 214-216°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty