Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Cyano-2-methoxyphenylboronic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

047126-500MG 047126-1G
101878-550EA 98.46 USD InStock
101878-550 101878-552
5-Cyano-2-methoxyphenylboronic acid ≥95%
5-Cyano-2-methoxyphenylboronic acid
Hazard: Irritant


Formula: C₈H₈BNO₃
Melting Pt: 211…212 °C
MDL Number: MFCD06801691
CAS Number: 612833-37-1
Order Now
Specification Test Results
Melting point 211-212°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty