Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-6-(1H-imidazol-1-yl)nicotinate ≥95%

Supplier: MATRIX SCIENTIFIC MS

047189-500MG 047189-1G 047189-5G
101878-852EA 181.54 USD InStock
101878-852 101878-854 101878-856
Ethyl-6-(1H-imidazol-1-yl)nicotinate ≥95%
Ethyl-6-(1H-imidazol-1-yl)nicotinate
Hazard: Irritant


Formula: C₁₁H₁₁N₃O₂
Melting Pt: 110…112 °C
MDL Number: MFCD12498710
CAS Number: 1171919-01-9
Order Now
Specification Test Results
Melting point 110-112°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty