Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenyl-N-[3-(dimethylamino)-2,2-dimethylpropyl]carbamate hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

047223-500MG 047223-5G
101879-000EA 181.54 USD InStock
101879-000 101879-004
Phenyl-N-[3-(dimethylamino)-2,2-dimethylpropyl]carbamate hydrochloride ≥95%
Phenyl-N-[3-(dimethylamino)-2,2-dimethylpropyl]carbamate hydrochloride
Hazard: Irritant


Formula: C₁₄H₂₃ClN₂O₂
Melting Pt: 156…158 °C
MDL Number: MFCD09027101
CAS Number: 1170396-21-0
Order Now
Specification Test Results
Melting point 156-158°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty