Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-[2-(3-formylphenoxy)ethoxy]benzenecarboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

047270-500MG 047270-5G
101879-216EA 278.46 USD InStock
101879-216 101879-220
Methyl-4-[2-(3-formylphenoxy)ethoxy]benzenecarboxylate ≥95%
Methyl-4-[2-(3-formylphenoxy)ethoxy]benzenecarboxylate
Hazard: Irritant


Formula: C₁₇H₁₆O₅
MW: 300.31 g/mol
Melting Pt: 115…117 °C
MDL Number: MFCD08689760
CAS Number: 937601-83-7
Order Now
Specification Test Results
Melting point 115-117°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty