Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Bromomethyl)-2,1,3-benzothiadiazole ≥95%

Supplier: MATRIX SCIENTIFIC MS

047299-500MG 047299-5G
101879-388EA 375.38 USD InStock
101879-388 101879-392
5-(Bromomethyl)-2,1,3-benzothiadiazole ≥95%
5-(Bromomethyl)-2,1,3-benzothiadiazole
Hazard: Irritant


Formula: C₇H₅BrN₂S
Melting Pt: 86-88°C
MDL Number: MFCD00544836
CAS Number: 65858-50-6
Order Now
Specification Test Results
Melting point 86-88°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty