Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-2-[(E)-(4-bromo-2-fluorophenyl)methylidene]-1-hydrazinecarboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

047529-500MG 047529-5G
101880-456EA 181.54 USD InStock
101880-456 101880-460
tert-Butyl-2-[(E)-(4-bromo-2-fluorophenyl)methylidene]-1-hydrazinecarboxylate ≥95%
tert-Butyl-2-[(E)-(4-bromo-2-fluorophenyl)methylidene]-1-hydrazinecarboxylate
Hazard: Irritant


Formula: C₁₂H₁₄BrFN₂O₂
Melting Pt: 182…184 °C
MDL Number: MFCD12025941
CAS Number: 1135283-46-3
Order Now
Specification Test Results
Melting point 182-184°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty