Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(5-Chloro-2-nitrophenyl)pyrrolidine ≥95%

Supplier: MATRIX SCIENTIFIC MS

047544-500MG 047544-1G 047544-5G
101880-514EA 375.38 USD InStock
101880-514 101880-516 101880-518
1-(5-Chloro-2-nitrophenyl)pyrrolidine ≥95%
1-(5-Chloro-2-nitrophenyl)pyrrolidine
Hazard: Irritant


Formula: C₁₀H₁₁ClN₂O₂
Melting Pt: 83…84 °C
CAS Number: 133387-30-1
Order Now
Specification Test Results
Melting point 83-84°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty