Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dibromo-2-methoxybenzenecarboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

047637-500MG 047637-1G 047637-5G
101880-822EA 49.23 USD InStock
101880-822 101880-824 101880-826
3,5-Dibromo-2-methoxybenzenecarboxylic acid ≥95%
3,5-Dibromo-2-methoxybenzenecarboxylic acid
Hazard: Irritant


Formula: C₈H₆Br₂O₃
Melting Pt: 198…200 °C
MDL Number: MFCD00016510
CAS Number: 13130-23-9
Order Now
Specification Test Results
Melting point 198-200°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty