Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dichloro-4-methoxybenzenecarboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

047639-500MG 047639-1G 047639-5G
101880-834EA 636.92 USD InStock
101880-834 101880-836 101880-838
3,5-Dichloro-4-methoxybenzenecarboxylic acid ≥95%
3,5-Dichloro-4-methoxybenzenecarboxylic acid
Hazard: Irritant


Formula: C₈H₆Cl₂O₃
MW: 221.04 g/mol
Melting Pt: 199…200 °C
MDL Number: MFCD00210404
CAS Number: 37908-97-7
Order Now
Specification Test Results
Melting point 199-200°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty