Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-{4-[(4-Fluorobenzyl)oxy]phenyl}acrylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

047641-500MG 047641-1G 047641-5G
101880-844EA 278.46 USD InStock
101880-844 101880-846 101880-848
3-{4-[(4-Fluorobenzyl)oxy]phenyl}acrylic acid ≥95%
3-{4-[(4-Fluorobenzyl)oxy]phenyl}acrylic acid
Hazard: Irritant


Formula: C₁₆H₁₃FO₃
MW: 272.28 g/mol
Melting Pt: 209…210 °C
MDL Number: MFCD00173647
CAS Number: 175136-19-3
Order Now
Specification Test Results
Melting point 209-210°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty