Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3-Chlorophenyl)-2-oxoethyl thiocyanate ≥95%

Supplier: MATRIX SCIENTIFIC MS

047725-500MG 047725-1G 047725-5G
101881-110EA 193.85 USD InStock
101881-110 101881-112 101881-114
2-(3-Chlorophenyl)-2-oxoethyl thiocyanate ≥95%
2-(3-Chlorophenyl)-2-oxoethyl thiocyanate
Hazard: Irritant


Formula: C₉H₆ClNOS
Melting Pt: 87…89 °C
MDL Number: MFCD09027128
CAS Number: 43045-12-1
Order Now
Specification Test Results
Melting point 87-89°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty