Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-[4-Bromo-2-nitro-5-(trifluoromethyl)phenyl]-2,2,2-trifluoroacetamide ≥95%

Supplier: MATRIX SCIENTIFIC MS

054487-500MG 054487-1G 054487-5G
101907-474EA 375.38 USD InStock
101907-474 101907-476 101907-478
N-[4-Bromo-2-nitro-5-(trifluoromethyl)phenyl]-2,2,2-trifluoroacetamide ≥95%
N-[4-Bromo-2-nitro-5-(trifluoromethyl)phenyl]-2,2,2-trifluoroacetamide
Hazard: Irritant


Formula: C₉H₃BrF₆N₂O₃
Melting Pt: 108…110 °C
MDL Number: MFCD14584783
CAS Number: 156425-51-3
Order Now
Specification Test Results
Melting point 108-110°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty