Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-4-(trifluoromethyl)nicotinic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

054510-500MG 054510-1G
101907-568EA 478.46 USD InStock
101907-568 101907-570
2-Methoxy-4-(trifluoromethyl)nicotinic acid ≥95%
2-Methoxy-4-(trifluoromethyl)nicotinic acid
Hazard: Irritant


Formula: C₈H₆F₃NO₃
MW: 221.14 g/mol
Melting Pt: 118…119 °C
MDL Number: MFCD14584806
CAS Number: 1221792-53-5
Order Now
Specification Test Results
Melting point 118-119°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty