Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-chloro-5-cyanopicolinate ≥95%

Supplier: MATRIX SCIENTIFIC MS

054522-500MG 054522-1G
101907-616EA 1109.23 USD InStock
101907-616 101907-618
Ethyl-3-chloro-5-cyanopicolinate ≥95%
Ethyl 3-chloro-5-cyanopicolinate
Hazard: Irritant


Formula: C₉H₇ClN₂O₂
Melting Pt: 51-52°C
MDL Number: MFCD14584814
CAS Number: 1221791-91-8
Order Now
Specification Test Results
Melting point 51-52°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty