Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-8-chloro-6-cyanoimidazo[1,2-a]pyridine-2-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

054538-500MG 054538-1G 054538-5G
101907-684EA 556.92 USD InStock
101907-684 101907-686 101907-688
Ethyl-8-chloro-6-cyanoimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 8-chloro-6-cyanoimidazo[1,2-a]pyridine-2-carboxylate
Hazard: Irritant


Formula: C₁₁H₈ClN₃O₂
Melting Pt: 189-190°C
MDL Number: MFCD14584828
CAS Number: 1221792-50-2
Order Now
Specification Test Results
Melting point 189-190°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty