Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1'-Benzyl-3H-spiro[1,3-benzothiazole-2,4'-piperidine] ≥95%

Supplier: MATRIX SCIENTIFIC MS

054684-500MG 054684-1G
101908-120EA 278.46 USD InStock
101908-120 101908-122
1'-Benzyl-3H-spiro[1,3-benzothiazole-2,4'-piperidine] ≥95%
1'-Benzyl-3H-spiro[1,3-benzothiazole-2,4'-piperidine]
Hazard: Irritant


Formula: C₁₈H₂₀N₂S
Melting Pt: 127…129 °C
MDL Number: MFCD03102266
CAS Number: 63208-95-7
Order Now
Specification Test Results
Melting point 127-129°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty