Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Chloro-6-cyanoimidazo[1,2-a]pyridine-2-carboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

054699-500MG 054699-1G
101908-182EA 507.69 USD InStock
101908-182 101908-184
8-Chloro-6-cyanoimidazo[1,2-a]pyridine-2-carboxylic acid ≥95%
8-Chloro-6-cyanoimidazo[1,2-a]pyridine-2-carboxylic acid
Hazard: Irritant


Formula: C₉H₄ClN₃O₂
Melting Pt: 242 °C (dec.)
MDL Number: MFCD16140274
CAS Number: 1221792-13-7
Order Now
Specification Test Results
Melting point 242°C(Dec)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty