Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Phenyl-3H-spiro[1,3-benzothiazole-2,1'-cyclohexane] ≥95%

Supplier: MATRIX SCIENTIFIC MS

054825-500MG 054825-1G
101908-684EA 278.46 USD InStock
101908-684 101908-686
4'-Phenyl-3H-spiro[1,3-benzothiazole-2,1'-cyclohexane] ≥95%
4'-Phenyl-3H-spiro[1,3-benzothiazole-2,1'-cyclohexane]
Hazard: Irritant


Formula: C₁₈H₁₉NS
Melting Pt: 110…111 °C
MDL Number: MFCD16140316
CAS Number: 121714-55-4
Order Now
Specification Test Results
Melting point 110-111°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty