Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-carbamimidamido-4-methyl-1,3-thiazole-5-carboxylate hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

059566-500MG 059566-1G 059566-5G
101918-460EA 193.85 USD InStock
101918-460 101918-462 101918-464
Ethyl-2-carbamimidamido-4-methyl-1,3-thiazole-5-carboxylate hydrochloride ≥95%
Ethyl-2-carbamimidamido-4-methyl-1,3-thiazole-5-carboxylate hydrochloride
Hazard: Irritant


Formula: C₈H₁₃ClN₄O₂S
Melting Pt: 219…221 °C
MDL Number: MFCD18651776
CAS Number: 131184-96-8
Order Now
Specification Test Results
Melting point 219-221°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty