Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-nitro-3-(piperidin-1-yl)benzoate ≥95%

Supplier: MATRIX SCIENTIFIC MS

059589-500MG 059589-1G 059589-5G
101918-574EA 403.08 USD InStock
101918-574 101918-576 101918-578
Methyl-4-nitro-3-(piperidin-1-yl)benzoate ≥95%
Methyl-4-nitro-3-(piperidin-1-yl)benzoate
Hazard: Irritant


Formula: C₁₃H₁₆N₂O₄
MW: 264.28 g/mol
Melting Pt: 95…97 °C
MDL Number: MFCD18651769
CAS Number: 1272756-51-0
Order Now
Specification Test Results
Melting point 95-97°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty