Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-4-[(2-aminobenzene)amido]piperidine-1-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

059594-500MG 059594-1G
101918-592EA 193.85 USD InStock
101918-592 101918-594
tert-Butyl-4-[(2-aminobenzene)amido]piperidine-1-carboxylate ≥95%
tert-Butyl-4-[(2-aminobenzene)amido]piperidine-1-carboxylate
Hazard: Irritant


Formula: C₁₇H₂₅N₃O₃
Melting Pt: 167…169 °C
MDL Number: MFCD08690356
CAS Number: 1021284-59-2
Order Now
Specification Test Results
Melting point 167-169°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty